Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGCATGAGTAAAATCTAGTTCTT[A/C]TGTTGGAGAAGGGTAATGTCTAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNPPD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CNPPD1 - cyclin Pas1/PHO80 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321389.1 | Intron | NP_001308318.1 | ||||
NM_001321390.1 | Intron | NP_001308319.1 | ||||
NM_001321391.1 | Intron | NP_001308320.1 | ||||
NM_015680.5 | Intron | NP_056495.3 |
FAM134A - family with sequence similarity 134 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321109.1 | Intron | NP_001308038.1 | ||||
NM_001321110.1 | Intron | NP_001308039.1 | ||||
NM_024293.5 | Intron | NP_077269.3 | ||||
XM_005246848.2 | Intron | XP_005246905.1 |
SLC23A3 - solute carrier family 23 member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |