Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGATTTGGACTATGCCTGCAGT[C/T]TGCTAGCACCAAGGGGACAGGCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604139 MIM: 601773 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAJB2 - DnaJ heat shock protein family (Hsp40) member B2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR153-1 - microRNA 153-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTPRN - protein tyrosine phosphatase, receptor type N | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199763.1 | Intron | NP_001186692.1 | ||||
NM_001199764.1 | Intron | NP_001186693.1 | ||||
NM_002846.3 | Intron | NP_002837.1 | ||||
XM_011511566.1 | Intron | XP_011509868.1 | ||||
XM_017004609.1 | Intron | XP_016860098.1 | ||||
XM_017004610.1 | Intron | XP_016860099.1 | ||||
XM_017004611.1 | Intron | XP_016860100.1 | ||||
XM_017004612.1 | Intron | XP_016860101.1 |