Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTTTCCTGAATTTCCCCACCTAT[C/T]ATAAACTCGTCCAGGATGAAATAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615781 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP1S3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AP1S3 - adaptor related protein complex 1 sigma 3 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039569.1 | 506 | Missense Mutation | ATA,ATG | I,M 118 | NP_001034658.1 | |
XM_011510600.2 | 506 | Intron | XP_011508902.1 |