Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAATATTCAGGATTTAAAGCAGCT[C/G]CTCTAAAGGCTACATTCACTGCAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RHBDD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RHBDD1 - rhomboid domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167608.1 | 175 | UTR 5 | NP_001161080.1 | |||
NM_032276.3 | 175 | Intron | NP_115652.2 | |||
XM_017005083.1 | 175 | UTR 5 | XP_016860572.1 | |||
XM_017005084.1 | 175 | UTR 5 | XP_016860573.1 | |||
XM_017005085.1 | 175 | Intron | XP_016860574.1 | |||
XM_017005086.1 | 175 | Intron | XP_016860575.1 | |||
XM_017005087.1 | 175 | Intron | XP_016860576.1 | |||
XM_017005088.1 | 175 | UTR 5 | XP_016860577.1 | |||
XM_017005089.1 | 175 | Intron | XP_016860578.1 | |||
XM_017005090.1 | 175 | UTR 5 | XP_016860579.1 | |||
XM_017005091.1 | 175 | UTR 5 | XP_016860580.1 | |||
XM_017005092.1 | 175 | Intron | XP_016860581.1 | |||
XM_017005093.1 | 175 | UTR 5 | XP_016860582.1 | |||
XM_017005094.1 | 175 | Intron | XP_016860583.1 | |||
XM_017005095.1 | 175 | UTR 5 | XP_016860584.1 |