Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAATGATACACATCGTTACTGTA[C/T]TTACTTATTGAGCACATACCGTATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600862 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGFG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AGFG1 - ArfGAP with FG repeats 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135187.1 | Intron | NP_001128659.1 | ||||
NM_001135188.1 | Intron | NP_001128660.1 | ||||
NM_001135189.1 | Intron | NP_001128661.1 | ||||
NM_004504.4 | Intron | NP_004495.2 | ||||
XM_005246516.3 | Intron | XP_005246573.1 | ||||
XM_006712478.3 | Intron | XP_006712541.1 | ||||
XM_006712479.3 | Intron | XP_006712542.1 | ||||
XM_006712480.3 | Intron | XP_006712543.1 | ||||
XM_006712481.3 | Intron | XP_006712544.1 | ||||
XM_017003954.1 | Intron | XP_016859443.1 |
MIR5703 - microRNA 5703 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |