Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTGCCCTAAATATGGGATAATAA[A/G]AATTTGGAGAATTGTATTGGAAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604457 MIM: 608602 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SP110 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SP110 - SP110 nuclear body protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001185015.1 | Intron | NP_001171944.1 | ||||
NM_004509.3 | Intron | NP_004500.3 | ||||
NM_004510.3 | Intron | NP_004501.3 | ||||
NM_080424.2 | Intron | NP_536349.2 | ||||
XM_005246525.3 | Intron | XP_005246582.1 | ||||
XM_006712487.3 | Intron | XP_006712550.1 | ||||
XM_006712489.3 | Intron | XP_006712552.1 | ||||
XM_011511088.2 | Intron | XP_011509390.1 | ||||
XM_011511089.2 | Intron | XP_011509391.1 | ||||
XM_011511090.2 | Intron | XP_011509392.1 | ||||
XM_011511091.2 | Intron | XP_011509393.1 | ||||
XM_011511092.2 | Intron | XP_011509394.1 | ||||
XM_017003968.1 | Intron | XP_016859457.1 | ||||
XM_017003969.1 | Intron | XP_016859458.1 | ||||
XM_017003970.1 | Intron | XP_016859459.1 |