Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602090 MIM: 607982 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LTBP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LTBP3 - latent transforming growth factor beta binding protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCYL1 - SCY1 like pseudokinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001048218.1 | Intron | NP_001041683.1 | ||||
NM_020680.3 | Intron | NP_065731.3 | ||||
XM_005274118.3 | Intron | XP_005274175.1 | ||||
XM_005274120.3 | Intron | XP_005274177.1 | ||||
XM_005274121.3 | Intron | XP_005274178.1 | ||||
XM_017018029.1 | Intron | XP_016873518.1 | ||||
XM_017018030.1 | Intron | XP_016873519.1 | ||||
XM_017018031.1 | Intron | XP_016873520.1 | ||||
XM_017018032.1 | Intron | XP_016873521.1 | ||||
XM_017018033.1 | Intron | XP_016873522.1 |
Set Membership: |
HapMap |