Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610110 MIM: 608566 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANO3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ANO3 - anoctamin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001313726.1 | Intron | NP_001300655.1 | ||||
NM_001313727.1 | Intron | NP_001300656.1 | ||||
NM_031418.3 | Intron | NP_113606.2 | ||||
XM_011520282.2 | Intron | XP_011518584.1 | ||||
XM_017018118.1 | Intron | XP_016873607.1 | ||||
XM_017018119.1 | Intron | XP_016873608.1 |
MUC15 - mucin 15, cell surface associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135091.1 | Intron | NP_001128563.1 | ||||
NM_001135092.1 | Intron | NP_001128564.1 | ||||
NM_145650.3 | Intron | NP_663625.2 | ||||
XM_017017235.1 | Intron | XP_016872724.1 | ||||
XM_017017236.1 | Intron | XP_016872725.1 |
Set Membership: |
HapMap Validated |