Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608925 MIM: 602998 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADIRF PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ADIRF - adipogenesis regulatory factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MMRN2 - multimerin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024756.2 | Intron | NP_079032.2 | ||||
XM_005270153.1 | Intron | XP_005270210.1 | ||||
XM_006717970.3 | Intron | XP_006718033.1 |
SNCG - synuclein gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003087.2 | Intron | NP_003078.2 | ||||
XM_011540105.2 | Intron | XP_011538407.1 |
Set Membership: |
HapMap |