Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
40 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608398 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSMD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CSMD2 - CUB and Sushi multiple domains 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281956.1 | 14344 | UTR 3 | NP_001268885.1 | |||
NM_052896.4 | 14344 | UTR 3 | NP_443128.2 | |||
XM_017000185.1 | 14344 | UTR 3 | XP_016855674.1 | |||
XM_017000186.1 | 14344 | UTR 3 | XP_016855675.1 | |||
XM_017000187.1 | 14344 | UTR 3 | XP_016855676.1 | |||
XM_017000188.1 | 14344 | UTR 3 | XP_016855677.1 | |||
XM_017000189.1 | 14344 | Intron | XP_016855678.1 | |||
XM_017000190.1 | 14344 | UTR 3 | XP_016855679.1 | |||
XM_017000191.1 | 14344 | Intron | XP_016855680.1 | |||
XM_017000192.1 | 14344 | Intron | XP_016855681.1 | |||
XM_017000193.1 | 14344 | Intron | XP_016855682.1 | |||
XM_017000194.1 | 14344 | UTR 3 | XP_016855683.1 |