Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBC1D12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TBC1D12 - TBC1 domain family member 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015188.1 | Intron | NP_056003.1 | ||||
XM_006717732.3 | Intron | XP_006717795.2 | ||||
XM_006717733.2 | Intron | XP_006717796.1 | ||||
XM_011539558.2 | Intron | XP_011537860.1 | ||||
XM_017015996.1 | Intron | XP_016871485.1 | ||||
XM_017015997.1 | Intron | XP_016871486.1 | ||||
XM_017015998.1 | Intron | XP_016871487.1 | ||||
XM_017015999.1 | Intron | XP_016871488.1 |
Set Membership: |
HapMap |