Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613568 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZMYM4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZMYM4 - zinc finger MYM-type containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005095.2 | Intron | NP_005086.2 | ||||
XM_005271328.4 | Intron | XP_005271385.1 | ||||
XM_005271331.2 | Intron | XP_005271388.1 | ||||
XM_011542421.2 | Intron | XP_011540723.1 | ||||
XM_011542423.2 | Intron | XP_011540725.1 | ||||
XM_011542425.2 | Intron | XP_011540727.1 | ||||
XM_017002803.1 | Intron | XP_016858292.1 | ||||
XM_017002804.1 | Intron | XP_016858293.1 | ||||
XM_017002805.1 | Intron | XP_016858294.1 |