Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATTCCAACACATTTCATCCTCCC[A/G]GCATTCATTTGTGGTCTCACACTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601548 | ||||||||||||||||||||
Literature Links: |
EFEMP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EFEMP1 - EGF containing fibulin like extracellular matrix protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039348.2 | 1493 | Missense Mutation | CGG,TGG | R,W 345 | NP_001034437.1 | |
NM_001039349.2 | 1493 | Missense Mutation | CGG,TGG | R,W 345 | NP_001034438.1 | |
XM_005264205.4 | 1493 | Missense Mutation | CGG,TGG | R,W 395 | XP_005264262.1 | |
XM_017003586.1 | 1493 | Missense Mutation | CGG,TGG | R,W 265 | XP_016859075.1 |