Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613297 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MARCH6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MARCH6 - membrane associated ring-CH-type finger 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270660.1 | Intron | NP_001257589.1 | ||||
NM_001270661.1 | Intron | NP_001257590.1 | ||||
NM_005885.3 | Intron | NP_005876.2 | ||||
XM_011513932.2 | Intron | XP_011512234.1 | ||||
XM_011513934.1 | Intron | XP_011512236.1 | ||||
XM_011513936.2 | Intron | XP_011512238.1 | ||||
XM_011513937.2 | Intron | XP_011512239.1 | ||||
XM_017008944.1 | Intron | XP_016864433.1 |