Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616642 MIM: 604209 | ||||||||||||||||||||
Literature Links: |
C6orf89 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C6orf89 - chromosome 6 open reading frame 89 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286635.1 | Intron | NP_001273564.1 | ||||
NM_001286636.1 | Intron | NP_001273565.1 | ||||
NM_001286637.1 | Intron | NP_001273566.1 | ||||
NM_152734.3 | Intron | NP_689947.2 | ||||
XM_011514377.2 | Intron | XP_011512679.1 | ||||
XM_017010432.1 | Intron | XP_016865921.1 | ||||
XM_017010433.1 | Intron | XP_016865922.1 | ||||
XM_017010434.1 | Intron | XP_016865923.1 |
CPNE5 - copine 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |