Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
40 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603504 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDC14A PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CDC14A - cell division cycle 14A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319210.1 | 264 | UTR 5 | NP_001306139.1 | |||
NM_001319211.1 | 264 | Intron | NP_001306140.1 | |||
NM_001319212.1 | 264 | UTR 5 | NP_001306141.1 | |||
NM_003672.3 | 264 | UTR 5 | NP_003663.2 | |||
NM_033312.2 | 264 | UTR 5 | NP_201569.1 | |||
NM_033313.2 | 264 | UTR 5 | NP_201570.1 | |||
XM_005271294.3 | 264 | Intron | XP_005271351.1 | |||
XM_005271296.3 | 264 | Intron | XP_005271353.1 | |||
XM_011542340.2 | 264 | Intron | XP_011540642.1 | |||
XM_011542341.2 | 264 | Intron | XP_011540643.1 | |||
XM_011542345.2 | 264 | Intron | XP_011540647.1 | |||
XM_011542346.2 | 264 | Intron | XP_011540648.1 | |||
XM_017002646.1 | 264 | Intron | XP_016858135.1 | |||
XM_017002647.1 | 264 | UTR 5 | XP_016858136.1 |
Set Membership: |
HapMap JSNP |