Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600285 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ETF1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
ETF1 - eukaryotic translation termination factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256302.1 | Intron | NP_001243231.1 | ||||
NM_001282185.1 | Intron | NP_001269114.1 | ||||
NM_001291974.1 | Intron | NP_001278903.1 | ||||
NM_001291975.1 | Intron | NP_001278904.1 | ||||
NM_004730.3 | Intron | NP_004721.1 | ||||
XM_005271921.1 | Intron | XP_005271978.1 | ||||
XM_017009221.1 | Intron | XP_016864710.1 | ||||
XM_017009222.1 | Intron | XP_016864711.1 |
Set Membership: |
HapMap |