Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCGGTGATCAGAAGCCTGACTCTG[G/T]AGCCTGACCCCATCATCGTTCCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613109 | ||||||||||||||||||||
Literature Links: |
GM2A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GM2A - GM2 ganglioside activator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000405.4 | 325 | Nonsense Mutation | GAG,TAG | E,* 54 | NP_000396.2 | |
NM_001167607.1 | 325 | Nonsense Mutation | GAG,TAG | E,* 54 | NP_001161079.1 |