Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
PRELID2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PRELID2 - PRELI domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138492.5 | Intron | NP_612501.3 | ||||
NM_182960.3 | Intron | NP_892005.1 | ||||
NM_205846.2 | Intron | NP_995318.1 | ||||
XM_017009127.1 | Intron | XP_016864616.1 | ||||
XM_017009128.1 | Intron | XP_016864617.1 | ||||
XM_017009129.1 | Intron | XP_016864618.1 | ||||
XM_017009130.1 | Intron | XP_016864619.1 | ||||
XM_017009131.1 | Intron | XP_016864620.1 | ||||
XM_017009132.1 | Intron | XP_016864621.1 | ||||
XM_017009133.1 | Intron | XP_016864622.1 | ||||
XM_017009134.1 | Intron | XP_016864623.1 | ||||
XM_017009135.1 | Intron | XP_016864624.1 |