Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600835 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CXCL12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CXCL12 - C-X-C motif chemokine ligand 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000609.6 | 2465 | UTR 3 | NP_000600.1 | |||
NM_001033886.2 | 2465 | Intron | NP_001029058.1 | |||
NM_001178134.1 | 2465 | Intron | NP_001171605.1 | |||
NM_001277990.1 | 2465 | UTR 3 | NP_001264919.1 | |||
NM_199168.3 | 2465 | Intron | NP_954637.1 |