Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605482 MIM: 612314 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GSTO1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GSTO1 - glutathione S-transferase omega 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GSTO2 - glutathione S-transferase omega 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001191013.1 | 25 | UTR 5 | NP_001177942.1 | |||
NM_001191014.1 | 25 | Intron | NP_001177943.1 | |||
NM_001191015.1 | 25 | Intron | NP_001177944.1 | |||
NM_183239.1 | 25 | UTR 5 | NP_899062.1 | |||
XM_006717624.3 | 25 | UTR 5 | XP_006717687.1 | |||
XM_011539270.2 | 25 | Intron | XP_011537572.1 | |||
XM_011539271.1 | 25 | UTR 5 | XP_011537573.1 | |||
XM_011539272.2 | 25 | UTR 5 | XP_011537574.1 | |||
XM_011539273.2 | 25 | UTR 5 | XP_011537575.1 | |||
XM_017015670.1 | 25 | UTR 5 | XP_016871159.1 | |||
XM_017015671.1 | 25 | UTR 5 | XP_016871160.1 |
MIR4482 - microRNA 4482 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |