Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164010 MIM: 615409 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHMP1A PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
CHMP1A - charged multivesicular body protein 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083314.3 | 118 | Intron | NP_001076783.1 | |||
NM_002768.4 | 118 | Intron | NP_002759.2 | |||
XM_011523098.1 | 118 | Intron | XP_011521400.1 |
SPATA33 - spermatogenesis associated 33 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271907.1 | 118 | Intron | NP_001258836.1 | |||
NM_001271908.1 | 118 | Intron | NP_001258837.1 | |||
NM_001271909.1 | 118 | UTR 5 | NP_001258838.1 | |||
NM_001271910.1 | 118 | Intron | NP_001258839.1 | |||
NM_153025.2 | 118 | Intron | NP_694570.1 | |||
XM_006721133.2 | 118 | Intron | XP_006721196.1 |
Set Membership: |
HapMap Validated |