Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 114240 MIM: 104180 | ||||||||||||||||||||
Literature Links: |
CAPN3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CAPN3 - calpain 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000070.2 | 535 | Missense Mutation | AAC,GAC | N,D 77 | NP_000061.1 | |
NM_024344.1 | 535 | Missense Mutation | AAC,GAC | N,D 77 | NP_077320.1 | |
NM_173087.1 | 535 | Missense Mutation | AAC,GAC | N,D 77 | NP_775110.1 | |
NM_173088.1 | 535 | Intron | NP_775111.1 | |||
NM_173089.1 | 535 | Intron | NP_775112.1 | |||
NM_173090.1 | 535 | Intron | NP_775113.1 |
GANC - glucosidase alpha, neutral C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |