Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C20orf197 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C20orf197 - chromosome 20 open reading frame 197 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302813.1 | 609 | Intron | NP_001289742.1 | |||
NM_001302815.1 | 609 | Intron | NP_001289744.1 | |||
NM_173644.2 | 609 | Intron | NP_775915.1 | |||
XM_017027828.1 | 609 | Intron | XP_016883317.1 | |||
XM_017027829.1 | 609 | UTR 5 | XP_016883318.1 | |||
XM_017027830.1 | 609 | UTR 5 | XP_016883319.1 | |||
XM_017027831.1 | 609 | Intron | XP_016883320.1 | |||
XM_017027832.1 | 609 | Intron | XP_016883321.1 |