Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACATGGACATTGTGCTAAGACTC[C/T]GAGCAGACTCCTGGAGGAACACCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180260 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100507291 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC100507291 - uncharacterized LOC100507291 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RBP1 - retinol binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130992.1 | Intron | NP_001124464.1 | ||||
NM_001130993.1 | Intron | NP_001124465.1 | ||||
NM_002899.3 | Intron | NP_002890.2 |