Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607951 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP57 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CEP57 - centrosomal protein 57 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243776.1 | 320 | Intron | NP_001230705.1 | |||
NM_001243777.1 | 320 | Intron | NP_001230706.1 | |||
NM_014679.4 | 320 | Intron | NP_055494.2 | |||
XM_006718945.2 | 320 | Intron | XP_006719008.1 | |||
XM_006718946.2 | 320 | Intron | XP_006719009.1 | |||
XM_017018592.1 | 320 | UTR 5 | XP_016874081.1 | |||
XM_017018593.1 | 320 | Intron | XP_016874082.1 | |||
XM_017018594.1 | 320 | Intron | XP_016874083.1 |
FAM76B - family with sequence similarity 76 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |