Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 102680 MIM: 602104 | ||||||||||||||||||||
Literature Links: |
ADD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADD1 - adducin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001119.4 | 676 | Intron | NP_001110.2 | |||
NM_001286645.1 | 676 | UTR 5 | NP_001273574.1 | |||
NM_014189.3 | 676 | Intron | NP_054908.2 | |||
NM_014190.3 | 676 | Intron | NP_054909.2 | |||
NM_176801.2 | 676 | Intron | NP_789771.1 | |||
XM_005247933.2 | 676 | Intron | XP_005247990.1 | |||
XM_005247934.2 | 676 | Intron | XP_005247991.1 | |||
XM_005247935.2 | 676 | Intron | XP_005247992.1 | |||
XM_005247936.2 | 676 | Intron | XP_005247993.1 | |||
XM_005247937.2 | 676 | Intron | XP_005247994.1 | |||
XM_017007703.1 | 676 | Intron | XP_016863192.1 | |||
XM_017007704.1 | 676 | Intron | XP_016863193.1 | |||
XM_017007705.1 | 676 | Intron | XP_016863194.1 | |||
XM_017007706.1 | 676 | Intron | XP_016863195.1 | |||
XM_017007707.1 | 676 | Intron | XP_016863196.1 | |||
XM_017007708.1 | 676 | Intron | XP_016863197.1 | |||
XM_017007709.1 | 676 | Intron | XP_016863198.1 | |||
XM_017007710.1 | 676 | Intron | XP_016863199.1 |
SH3BP2 - SH3 domain binding protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |