Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACTGGCCTAGTCCCCAAGGCTGA[A/C]GAAACATGGTCTGGCCTGACCCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616254 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLPB PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CLPB - ClpB homolog, mitochondrial AAA ATPase chaperonin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258392.2 | 2978 | UTR 3 | NP_001245321.1 | |||
NM_001258393.2 | 2978 | UTR 3 | NP_001245322.1 | |||
NM_001258394.2 | 2978 | UTR 3 | NP_001245323.1 | |||
NM_030813.5 | 2978 | UTR 3 | NP_110440.1 | |||
XM_005274320.1 | 2978 | Intron | XP_005274377.1 | |||
XM_011545289.2 | 2978 | Intron | XP_011543591.1 |
LOC105369375 - uncharacterized LOC105369375 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |