Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCCAGCAAAGGTGGCGCGGAGGC[A/G]GGCCACAGCCTCAGGCTGGCCAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605832 MIM: 601002 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACSS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACSS2 - acyl-CoA synthetase short-chain family member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GSS - glutathione synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000178.3 | 1087 | Missense Mutation | CGC,TGC | R,C 330 | NP_000169.1 | |
NM_001322494.1 | 1087 | Missense Mutation | CGC,TGC | R,C 330 | NP_001309423.1 | |
NM_001322495.1 | 1087 | Missense Mutation | CGC,TGC | R,C 330 | NP_001309424.1 |