Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603080 MIM: 615097 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC6A12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC6A12 - solute carrier family 6 member 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC6A13 - solute carrier family 6 member 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190997.2 | Intron | NP_001177926.1 | ||||
NM_001243392.1 | Intron | NP_001230321.1 | ||||
NM_016615.4 | Intron | NP_057699.2 | ||||
XM_006719008.3 | Intron | XP_006719071.1 | ||||
XM_011521012.2 | Intron | XP_011519314.1 | ||||
XM_011521013.1 | Intron | XP_011519315.1 | ||||
XM_011521014.1 | Intron | XP_011519316.1 | ||||
XM_017019842.1 | Intron | XP_016875331.1 | ||||
XM_017019843.1 | Intron | XP_016875332.1 | ||||
XM_017019844.1 | Intron | XP_016875333.1 | ||||
XM_017019845.1 | Intron | XP_016875334.1 | ||||
XM_017019846.1 | Intron | XP_016875335.1 | ||||
XM_017019847.1 | Intron | XP_016875336.1 |
Set Membership: |
HapMap |