Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605629 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CIT PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CIT - citron rho-interacting serine/threonine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206999.1 | Intron | NP_001193928.1 | ||||
NM_007174.2 | Intron | NP_009105.1 | ||||
XM_006719206.2 | Intron | XP_006719269.1 | ||||
XM_011537783.1 | Intron | XP_011536085.1 | ||||
XM_011537784.1 | Intron | XP_011536086.1 | ||||
XM_011537785.1 | Intron | XP_011536087.1 | ||||
XM_011537787.1 | Intron | XP_011536089.1 | ||||
XM_011537788.1 | Intron | XP_011536090.1 | ||||
XM_011537789.1 | Intron | XP_011536091.1 | ||||
XM_011537790.2 | Intron | XP_011536092.1 | ||||
XM_011537791.2 | Intron | XP_011536093.1 | ||||
XM_011537792.2 | Intron | XP_011536094.1 | ||||
XM_017018735.1 | Intron | XP_016874224.1 | ||||
XM_017018736.1 | Intron | XP_016874225.1 | ||||
XM_017018737.1 | Intron | XP_016874226.1 | ||||
XM_017018738.1 | Intron | XP_016874227.1 | ||||
XM_017018739.1 | Intron | XP_016874228.1 |
Set Membership: |
HapMap Validated |