Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607749 MIM: 139130 MIM: 610342 MIM: 601447 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDCA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global - Not Available | Caucasian
|
CEPH (CEU)
|
||||||
EAS - Not Available | African American
|
YRI (Yoruba)
|
||||||
SAS - Not Available | Japanese
|
JPT (Japanese)
|
||||||
AFR - Not Available | Chinese
|
CHB (Han Chinese)
|
||||||
EUR - Not Available | ||||||||
AMR - Not Available |
CDCA3 - cell division cycle associated 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297602.1 | 1351 | Intron | NP_001284531.1 | |||
NM_001297603.1 | 1351 | Intron | NP_001284532.1 | |||
NM_001297604.1 | 1351 | Intron | NP_001284533.1 | |||
NM_031299.5 | 1351 | Intron | NP_112589.1 |
GNB3 - G protein subunit beta 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297571.1 | 1351 | UTR 3 | NP_001284500.1 | |||
NM_002075.3 | 1351 | UTR 3 | NP_002066.1 | |||
XM_011520953.2 | 1351 | UTR 3 | XP_011519255.1 |
P3H3 - prolyl 3-hydroxylase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP5 - ubiquitin specific peptidase 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |