Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607252 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
APOL2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
APOL2 - apolipoprotein L2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030882.3 | 1210 | Intron | NP_112092.2 | |||
NM_145637.2 | 1210 | Missense Mutation | ATC,GTC | I,V 245 | NP_663612.2 | |
XM_011530074.1 | 1210 | Intron | XP_011528376.1 | |||
XM_011530075.1 | 1210 | Intron | XP_011528377.1 | |||
XM_011530076.2 | 1210 | Intron | XP_011528378.1 | |||
XM_011530077.2 | 1210 | Intron | XP_011528379.1 | |||
XM_011530078.1 | 1210 | Intron | XP_011528380.1 | |||
XM_017028724.1 | 1210 | Intron | XP_016884213.1 |
Set Membership: |
HapMap |