Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606272 MIM: 605060 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTNS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTNS - cystinosin, lysosomal cystine transporter | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031681.2 | Intron | NP_001026851.2 | ||||
NM_004937.2 | Intron | NP_004928.2 | ||||
XM_005256485.2 | Intron | XP_005256542.1 | ||||
XM_006721463.2 | Intron | XP_006721526.1 | ||||
XM_006721464.2 | Intron | XP_006721527.1 | ||||
XM_011523691.2 | Intron | XP_011521993.1 | ||||
XM_011523692.2 | Intron | XP_011521994.1 | ||||
XM_017024254.1 | Intron | XP_016879743.1 | ||||
XM_017024255.1 | Intron | XP_016879744.1 | ||||
XM_017024256.1 | Intron | XP_016879745.1 | ||||
XM_017024257.1 | Intron | XP_016879746.1 | ||||
XM_017024258.1 | Intron | XP_016879747.1 |
SHPK - sedoheptulokinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |