Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGGCAGGGTGCTACTTAAGCCTC[A/G]ATGGTACCATTAACTCCACGTAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607909 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AZIN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
AZIN1 - antizyme inhibitor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301668.1 | Intron | NP_001288597.1 | ||||
NM_015878.5 | Intron | NP_056962.2 | ||||
NM_148174.3 | Intron | NP_680479.1 | ||||
XM_005250969.1 | Intron | XP_005251026.1 | ||||
XM_011517126.1 | Intron | XP_011515428.1 | ||||
XM_011517127.1 | Intron | XP_011515429.1 | ||||
XM_011517128.2 | Intron | XP_011515430.1 | ||||
XM_017013563.1 | Intron | XP_016869052.1 | ||||
XM_017013564.1 | Intron | XP_016869053.1 | ||||
XM_017013565.1 | Intron | XP_016869054.1 | ||||
XM_017013566.1 | Intron | XP_016869055.1 |
LOC107986873 - forkhead box protein L2-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017014121.1 | Intron | XP_016869610.1 |
Set Membership: |
HapMap Validated |