Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTGGCCCTGGAGCATGTGGGCCC[C/T]GGGGCGTGGGTTGAGCTCGGTCTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610312 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100507071 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC100507071 - uncharacterized LOC100507071 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIWIL2 - piwi like RNA-mediated gene silencing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135721.1 | 9 | Intron | NP_001129193.1 | |||
NM_018068.3 | 9 | UTR 5 | NP_060538.2 | |||
XM_005273550.4 | 9 | UTR 5 | XP_005273607.1 | |||
XM_005273551.3 | 9 | UTR 5 | XP_005273608.1 |
Set Membership: |
HapMap |