Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCTGAATGATTTTTCTTTACAAG[C/T]TGGAAGTTTAGGAAAGCTCGAATTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123842 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPIC PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PPIC - peptidylprolyl isomerase C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |