Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615904 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PXDNL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PXDNL - peroxidasin like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144651.4 | Intron | NP_653252.3 | ||||
XM_005251168.3 | Intron | XP_005251225.1 | ||||
XM_006716420.3 | Intron | XP_006716483.1 | ||||
XM_011517456.2 | Intron | XP_011515758.1 | ||||
XM_011517457.2 | Intron | XP_011515759.1 | ||||
XM_011517458.2 | Intron | XP_011515760.1 | ||||
XM_011517459.2 | Intron | XP_011515761.1 | ||||
XM_017013041.1 | Intron | XP_016868530.1 |