Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615662 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SERPINB12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SERPINB12 - serpin family B member 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001307928.1 | Intron | NP_001294857.1 | ||||
NM_080474.2 | Intron | NP_536722.1 | ||||
XM_005266778.3 | Intron | XP_005266835.2 | ||||
XM_011526246.1 | Intron | XP_011524548.1 | ||||
XM_011526248.1 | Intron | XP_011524550.1 | ||||
XM_011526249.1 | Intron | XP_011524551.1 | ||||
XM_017026067.1 | Intron | XP_016881556.1 |