Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGAGATAAAAGTGAGAAGAGTCA[A/G]AAAGATTGATTTCTATAGGCCGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613183 MIM: 609281 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BOLA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BOLA3 - bolA family member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
BOLA3-AS1 - BOLA3 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MOB1A - MOB kinase activator 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317110.1 | 3647 | UTR 3 | NP_001304039.1 | |||
NM_001317111.1 | 3647 | UTR 3 | NP_001304040.1 | |||
NM_001317112.1 | 3647 | UTR 3 | NP_001304041.1 | |||
NM_018221.4 | 3647 | UTR 3 | NP_060691.2 | |||
XM_011532948.2 | 3647 | UTR 3 | XP_011531250.1 |
Set Membership: |
HapMap |