Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGATGCCCACCAGCGATTCAGGG[C/T]TCCCCGGAGAACAGAGAGGGGCGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601801 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC102724532 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC102724532 - uncharacterized LOC102724532 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SP2 - Sp2 transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003110.5 | 70 | Intron | NP_003101.3 | |||
XM_006722023.2 | 70 | Intron | XP_006722086.1 | |||
XM_006722025.2 | 70 | Intron | XP_006722088.1 | |||
XM_006722026.2 | 70 | Intron | XP_006722089.1 | |||
XM_006722027.2 | 70 | Intron | XP_006722090.1 | |||
XM_011525136.2 | 70 | Intron | XP_011523438.1 | |||
XM_011525137.2 | 70 | Intron | XP_011523439.1 | |||
XM_011525138.2 | 70 | Intron | XP_011523440.1 | |||
XM_011525139.2 | 70 | Intron | XP_011523441.1 | |||
XM_011525140.2 | 70 | Intron | XP_011523442.1 | |||
XM_011525142.1 | 70 | Intron | XP_011523444.1 | |||
XM_011525143.1 | 70 | Intron | XP_011523445.1 | |||
XM_017024968.1 | 70 | UTR 5 | XP_016880457.1 | |||
XM_017024969.1 | 70 | Intron | XP_016880458.1 |
SP2-AS1 - SP2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |