Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTTCTCTTTTTATAATTAAAACT[G/T]CTTCAAACAGTGGCTTCACTAGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615358 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AK9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AK9 - adenylate kinase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145128.2 | Intron | NP_001138600.2 | ||||
NM_145025.4 | Intron | NP_659462.1 | ||||
XM_006715376.3 | Intron | XP_006715439.2 | ||||
XM_011535550.2 | Intron | XP_011533852.1 | ||||
XM_011535552.2 | Intron | XP_011533854.1 | ||||
XM_011535553.2 | Intron | XP_011533855.1 | ||||
XM_011535554.2 | Intron | XP_011533856.1 | ||||
XM_011535555.2 | Intron | XP_011533857.1 | ||||
XM_011535556.2 | Intron | XP_011533858.1 | ||||
XM_011535558.1 | Intron | XP_011533860.1 | ||||
XM_011535559.2 | Intron | XP_011533861.1 | ||||
XM_017010384.1 | Intron | XP_016865873.1 | ||||
XM_017010385.1 | Intron | XP_016865874.1 | ||||
XM_017010386.1 | Intron | XP_016865875.1 | ||||
XM_017010387.1 | Intron | XP_016865876.1 | ||||
XM_017010388.1 | Intron | XP_016865877.1 | ||||
XM_017010389.1 | Intron | XP_016865878.1 |
Set Membership: |
HapMap |