Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGAGGGTTTTTGTATCTATCCCA[A/C]CCCCAAAAGCAGTGAATTACTGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBC1D19 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TBC1D19 - TBC1 domain family member 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001292054.1 | Intron | NP_001278983.1 | ||||
NM_018317.3 | Intron | NP_060787.2 | ||||
XM_011513852.2 | Intron | XP_011512154.1 | ||||
XM_017008363.1 | Intron | XP_016863852.1 | ||||
XM_017008364.1 | Intron | XP_016863853.1 |