Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609235 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRSK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BRSK1 - BR serine/threonine kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM150B - transmembrane protein 150B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085488.2 | Intron | NP_001078957.1 | ||||
NM_001282011.1 | Intron | NP_001268940.1 | ||||
XM_005258812.3 | Intron | XP_005258869.1 | ||||
XM_011526850.2 | Intron | XP_011525152.1 | ||||
XM_011526851.2 | Intron | XP_011525153.1 | ||||
XM_011526852.2 | Intron | XP_011525154.1 | ||||
XM_011526853.2 | Intron | XP_011525155.1 | ||||
XM_011526854.2 | Intron | XP_011525156.1 | ||||
XM_011526855.2 | Intron | XP_011525157.1 | ||||
XM_011526856.2 | Intron | XP_011525158.1 | ||||
XM_011526857.2 | Intron | XP_011525159.1 | ||||
XM_017026667.1 | Intron | XP_016882156.1 | ||||
XM_017026668.1 | Intron | XP_016882157.1 |