Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTTTTAACTAAGGGATGGTAAAT[A/G]GCATGGGCTTTATCCTGGATGCCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
48 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605596 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SF3A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SF3A3 - splicing factor 3a subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320830.1 | Intron | NP_001307759.1 | ||||
NM_006802.3 | Intron | NP_006793.1 |
Set Membership: |
Validated |