Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610924 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBCK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RBCK1 - RANBP2-type and C3HC4-type zinc finger containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323956.1 | 990 | Intron | NP_001310885.1 | |||
NM_001323958.1 | 990 | Intron | NP_001310887.1 | |||
NM_001323960.1 | 990 | Silent Mutation | TAC,TAT | Y,Y 94 | NP_001310889.1 | |
NM_006462.5 | 990 | Intron | NP_006453.1 | |||
NM_031229.3 | 990 | Intron | NP_112506.2 | |||
XM_005260645.1 | 990 | Intron | XP_005260702.1 | |||
XM_011529137.1 | 990 | Intron | XP_011527439.1 | |||
XM_011529138.1 | 990 | Intron | XP_011527440.1 | |||
XM_011529139.2 | 990 | Intron | XP_011527441.1 | |||
XM_011529140.1 | 990 | Intron | XP_011527442.1 | |||
XM_017027594.1 | 990 | Intron | XP_016883083.1 | |||
XM_017027595.1 | 990 | Intron | XP_016883084.1 |
Set Membership: |
HapMap |