Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACTGGGGCCTCACTCTCTCCTGG[C/T]TGGTCAGGAGCCCTGCCCAAAGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602618 MIM: 605918 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CTBP1 - C-terminal binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012614.1 | Intron | NP_001012632.1 | ||||
NM_001328.2 | Intron | NP_001319.1 | ||||
XM_005272261.3 | Intron | XP_005272318.1 | ||||
XM_005272263.4 | Intron | XP_005272320.1 | ||||
XM_011513395.2 | Intron | XP_011511697.1 | ||||
XM_011513396.2 | Intron | XP_011511698.1 | ||||
XM_011513397.2 | Intron | XP_011511699.1 | ||||
XM_017007762.1 | Intron | XP_016863251.1 | ||||
XM_017007763.1 | Intron | XP_016863252.1 | ||||
XM_017007764.1 | Intron | XP_016863253.1 | ||||
XM_017007765.1 | Intron | XP_016863254.1 | ||||
XM_017007766.1 | Intron | XP_016863255.1 | ||||
XM_017007767.1 | Intron | XP_016863256.1 |
CTBP1-AS - CTBP1 antisense RNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC100130872 - uncharacterized LOC100130872 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPON2 - spondin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
Validated |