Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102595 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACYP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ACYP2 - acylphosphatase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320586.1 | 1818 | Intron | NP_001307515.1 | |||
NM_001320587.1 | 1818 | Intron | NP_001307516.1 | |||
NM_001320588.1 | 1818 | Intron | NP_001307517.1 | |||
NM_001320589.1 | 1818 | Intron | NP_001307518.1 | |||
NM_001320590.1 | 1818 | Intron | NP_001307519.1 | |||
NM_138448.3 | 1818 | Intron | NP_612457.1 | |||
XM_017005406.1 | 1818 | Intron | XP_016860895.1 | |||
XM_017005407.1 | 1818 | Intron | XP_016860896.1 | |||
XM_017005408.1 | 1818 | Intron | XP_016860897.1 | |||
XM_017005409.1 | 1818 | Intron | XP_016860898.1 | |||
XM_017005410.1 | 1818 | Intron | XP_016860899.1 | |||
XM_017005411.1 | 1818 | Intron | XP_016860900.1 | |||
XM_017005412.1 | 1818 | Intron | XP_016860901.1 | |||
XM_017005413.1 | 1818 | Intron | XP_016860902.1 |
TSPYL6 - TSPY like 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003937.2 | 1818 | UTR 3 | NP_001003937.2 |
Set Membership: |
HapMap Validated |