Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTATTCACTTTTCAGTTCTCTTTG[A/T]CTTTTTGGTTGCTCTCAGATGAAGG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 603922 | |||||||||||||||||||||||
Literature Links: |
SUCLG2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
SUCLG2 - succinate-CoA ligase GDP-forming beta subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177599.1 | 1562 | Intron | NP_001171070.1 | |||
NM_003848.3 | 1562 | Intron | NP_003839.2 | |||
XM_017007415.1 | 1562 | Intron | XP_016862904.1 | |||
XM_017007416.1 | 1562 | Intron | XP_016862905.1 | |||
XM_017007417.1 | 1562 | Intron | XP_016862906.1 | |||
XM_017007418.1 | 1562 | Intron | XP_016862907.1 | |||
XM_017007419.1 | 1562 | UTR 3 | XP_016862908.1 | |||
XM_017007420.1 | 1562 | Intron | XP_016862909.1 |