Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
56 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601580 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAPZA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CAPZA1 - capping actin protein of muscle Z-line alpha subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006135.2 | Intron | NP_006126.1 | ||||
XM_011542225.2 | Intron | XP_011540527.1 | ||||
XM_017002424.1 | Intron | XP_016857913.1 |
ST7L - suppression of tumorigenicity 7 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308264.1 | Intron | NP_001295193.1 | ||||
NM_017744.4 | Intron | NP_060214.2 | ||||
NM_138727.3 | Intron | NP_620055.1 | ||||
NM_138728.2 | Intron | NP_620056.1 | ||||
NM_138729.3 | Intron | NP_620057.1 | ||||
XM_005270963.4 | Intron | XP_005271020.1 | ||||
XM_005270964.3 | Intron | XP_005271021.1 | ||||
XM_011541627.2 | Intron | XP_011539929.1 | ||||
XM_011541628.2 | Intron | XP_011539930.1 | ||||
XM_011541629.2 | Intron | XP_011539931.1 | ||||
XM_011541630.2 | Intron | XP_011539932.1 | ||||
XM_011541631.2 | Intron | XP_011539933.1 | ||||
XM_017001534.1 | Intron | XP_016857023.1 | ||||
XM_017001535.1 | Intron | XP_016857024.1 | ||||
XM_017001536.1 | Intron | XP_016857025.1 | ||||
XM_017001537.1 | Intron | XP_016857026.1 | ||||
XM_017001538.1 | Intron | XP_016857027.1 |
Set Membership: |
HapMap Validated |